Product Details

SNP ID
rs199752828
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:37863001 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTGTCAATGTAGTCTTCCAGCTCG[C/T]GGACCTGGAACTCCTTCTTGCTTAT
Phenotype
MIM: 608737
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RAB11FIP1 PubMed Links

Gene Details

Gene
RAB11FIP1
Gene Name
RAB11 family interacting protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001002814.2 3802 Missense Mutation CAC,CGC H1249R NP_001002814.2
NM_025151.4 3802 Missense Mutation CAC,CGC H615R NP_079427.4
XM_017013869.1 3802 Intron XP_016869358.1

View Full Product Details