Product Details

SNP ID
rs200284600
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:41646602 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGGTGCGCGGTCCCCGCCTGCGCC[C/T]GCACCCGCGCCGCCCGGGGCCCGGG
Phenotype
MIM: 612641 MIM: 610772
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ANK1 PubMed Links

Gene Details

Gene
ANK1
Gene Name
ankyrin 1
There are no transcripts associated with this gene.

Gene
NKX6-3
Gene Name
NK6 homeobox 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152568.3 259 Silent Mutation GCA,GCG A85A NP_689781.1
XM_017013143.1 259 Silent Mutation GCA,GCG A215A XP_016868632.1

View Full Product Details