Product Details

SNP ID
rs199498627
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:80487771 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTACCGAGCACAAACTCCACGAA[A/G]CCAACGCCCAGGTACAGTATATCCT
Phenotype
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ZBTB10 PubMed Links

Gene Details

Gene
ZBTB10
Gene Name
zinc finger and BTB domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001105539.2 1559 Missense Mutation ACC,GCC T321A NP_001099009.1
NM_001277145.1 1559 Intron NP_001264074.1
NM_023929.4 1559 Missense Mutation ACC,GCC T321A NP_076418.3

View Full Product Details