Product Details

SNP ID
rs199542428
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:91070346 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGAAGGTGCCTACTAGCCGGTGC[A/G]GGTTTCTTCTAGCGCGTGTGCTGGG
Phenotype
MIM: 612021
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OTUD6B PubMed Links

Gene Details

Gene
OTUD6B
Gene Name
OTU domain containing 6B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286745.1 42 UTR 5 NP_001273674.1
NM_016023.3 42 Silent Mutation AGG,GGG R18G NP_057107.3
XM_011517129.2 42 UTR 5 XP_011515431.2
Gene
OTUD6B-AS1
Gene Name
OTUD6B antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details