Product Details

SNP ID
rs199902693
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:141357003 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGTGCTGGCCGAAGAGGCCTCGGAC[A/G]GTGGCCTGGAAGCCACTGGTGACGA
Phenotype
MIM: 601908
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR20 PubMed Links

Gene Details

Gene
GPR20
Gene Name
G protein-coupled receptor 20
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005293.2 1010 Silent Mutation ACC,ACT T307T NP_005284.2

View Full Product Details