Product Details

SNP ID
rs200032631
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:66430038 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGACCAGGAGGAGGCCGCCAAG[A/T]GGAGGAAAATGAGCCAGAAGGGCAA
Phenotype
MIM: 611083
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ADHFE1 PubMed Links

Gene Details

Gene
ADHFE1
Gene Name
alcohol dehydrogenase, iron containing 1
There are no transcripts associated with this gene.

Gene
RRS1
Gene Name
ribosome biogenesis regulator homolog
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015169.3 1011 Missense Mutation AGG,TGG R303W NP_055984.1
Gene
RRS1-AS1
Gene Name
RRS1 antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details