Product Details

SNP ID
rs200074208
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:98949225 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCCTTCCCGGCCCTGCCTTTTAC[C/T]ACCCACCTATTCCACCCCAAGCAGG
Phenotype
MIM: 611297 MIM: 605030
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OSR2 PubMed Links

Gene Details

Gene
OSR2
Gene Name
odd-skipped related transciption factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142462.2 1016 Silent Mutation ACC,ACT T91T NP_001135934.1
NM_001286841.1 1016 Silent Mutation ACC,ACT T212T NP_001273770.1
NM_053001.3 1016 Silent Mutation ACC,ACT T91T NP_443727.2
XM_005250778.4 1016 Silent Mutation ACC,ACT T212T XP_005250835.1
XM_011516825.2 1016 Silent Mutation ACC,ACT T148T XP_011515127.1
XM_011516826.2 1016 Silent Mutation ACC,ACT T144T XP_011515128.1
XM_011516827.2 1016 Silent Mutation ACC,ACT T91T XP_011515129.1
XM_017013018.1 1016 Silent Mutation ACC,ACT T91T XP_016868507.1
Gene
STK3
Gene Name
serine/threonine kinase 3
There are no transcripts associated with this gene.

View Full Product Details