Product Details

SNP ID
hCV192028397
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144355996 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGTGCTTGGTGTGACCCGGGTGC[A/C]CCTGAGGTTAAGAGAGCACAGGGCT
Phenotype
MIM: 609076 MIM: 607882
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FBXL6 PubMed Links

Gene Details

Gene
FBXL6
Gene Name
F-box and leucine rich repeat protein 6
There are no transcripts associated with this gene.

Gene
SLC52A2
Gene Name
solute carrier family 52 member 2
There are no transcripts associated with this gene.

Gene
TMEM249
Gene Name
transmembrane protein 249
There are no transcripts associated with this gene.

View Full Product Details