Product Details

SNP ID
rs199865032
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:22691013 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTCCATGCCCTGGAAGGGCTTGTG[A/C]TCCGGAATGGAGCCCATGTCGTTGG
Phenotype
MIM: 602419
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
EGR3 PubMed Links

Gene Details

Gene
EGR3
Gene Name
early growth response 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199880.1 933 Missense Mutation GAG,GAT E170D NP_001186809.1
NM_001199881.1 933 Missense Mutation GAG,GAT E154D NP_001186810.1
NM_004430.2 933 Missense Mutation GAG,GAT E208D NP_004421.2
XM_005273425.3 933 Missense Mutation GAG,GAT E263D XP_005273482.1
XM_005273426.3 933 Missense Mutation GAG,GAT E170D XP_005273483.1
XM_011544429.2 933 Missense Mutation GAG,GAT E227D XP_011542731.1

View Full Product Details