Product Details
- SNP ID
-
rs199865645
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:140515197 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAAAGAGGAAAAGAGGGAAGAAGAT[A/G]AGGAGAATGACAATGATAATGAAAG
- Phenotype
-
MIM: 606229
MIM: 607268
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AGO2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2231524] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AGO2
- Gene Name
- argonaute 2, RISC catalytic component
There are no transcripts associated with this gene.
- Gene
- CHRAC1
- Gene Name
- chromatin accessibility complex 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_017444.5 |
548 |
Missense Mutation |
AAG,GAG |
K116E |
NP_059140.1 |
View Full Product Details