Product Details
- SNP ID
-
rs199654680
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.8:132572415 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGGTGTGTGTTTTTTCTCTTGAACT[A/G]TGTTAGTCACATCAGGGAATGAGTG
- Phenotype
-
MIM: 614930
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
LRRC6
PubMed Links
Gene Details
- Gene
- LRRC6
- Gene Name
- leucine rich repeat containing 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001321961.1 |
2049 |
Missense Mutation |
ACA,ATA |
T411I |
NP_001308890.1 |
NM_001321962.1 |
2049 |
Missense Mutation |
ACA,ATA |
T349I |
NP_001308891.1 |
NM_001321963.1 |
2049 |
Missense Mutation |
ACA,ATA |
T311I |
NP_001308892.1 |
NM_001321964.1 |
2049 |
Missense Mutation |
ACA,ATA |
T311I |
NP_001308893.1 |
NM_001321965.1 |
2049 |
Missense Mutation |
ACA,ATA |
T311I |
NP_001308894.1 |
NM_001321966.1 |
2049 |
Missense Mutation |
ACA,ATA |
T291I |
NP_001308895.1 |
NM_012472.5 |
2049 |
Missense Mutation |
ACA,ATA |
T431I |
NP_036604.2 |
XM_006716538.3 |
2049 |
Missense Mutation |
ACA,ATA |
T437I |
XP_006716601.2 |
XM_011516950.2 |
2049 |
Missense Mutation |
ACA,ATA |
T417I |
XP_011515252.1 |
XM_017013296.1 |
2049 |
Missense Mutation |
ACA,ATA |
T397I |
XP_016868785.1 |
XM_017013297.1 |
2049 |
Missense Mutation |
ACA,ATA |
T311I |
XP_016868786.1 |
XM_017013298.1 |
2049 |
Missense Mutation |
ACA,ATA |
T311I |
XP_016868787.1 |
View Full Product Details