Product Details

SNP ID
rs200200680
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128063543 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCGGAAATCTTTGATCATAGGC[C/T]GGAGGACCTGGATGAGGACGCTCAG
Phenotype
MIM: 610673 MIM: 608745
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NAIF1 PubMed Links

Gene Details

Gene
NAIF1
Gene Name
nuclear apoptosis inducing factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_197956.3 1088 Missense Mutation CAG,CGG Q290R NP_931045.1
Gene
SLC25A25
Gene Name
solute carrier family 25 member 25
There are no transcripts associated with this gene.

View Full Product Details