Product Details

SNP ID
rs200937901
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:101399912 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCTGACATTTGAGGATGTGGCTG[G/T]GTTTTTTACCCAGGAGGAGTGGGAT
Phenotype
MIM: 611854 MIM: 603132
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
MRPL50 PubMed Links

Gene Details

Gene
MRPL50
Gene Name
mitochondrial ribosomal protein L50
There are no transcripts associated with this gene.

Gene
ZNF189
Gene Name
zinc finger protein 189
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278231.1 516 Intron NP_001265160.1
NM_001278232.1 516 Intron NP_001265161.1
NM_001278240.1 516 Intron NP_001265169.1
NM_003452.3 516 Missense Mutation GGG,GTG G21V NP_003443.2
NM_197977.2 516 Intron NP_932094.1
XM_006717280.3 516 UTR 5 XP_006717343.1
XM_006717281.3 516 Intron XP_006717344.1
XM_011518998.2 516 Intron XP_011517300.1
XM_011518999.2 516 Intron XP_011517301.1
XM_017015121.1 516 Intron XP_016870610.1

View Full Product Details