Product Details
- SNP ID
-
rs201393222
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:95876027 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TTACATGCAGCCGGGCTCGGCCCCT[A/C]CCCCTGGCCGGATGGATCCGTCGGC
- Phenotype
-
MIM: 615667
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
ERCC6L2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs56108623] are located under a probe and SNP(s) [rs690528] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ERCC6L2
- Gene Name
- ERCC excision repair 6 like 2
- Gene
- LINC00476
- Gene Name
- long intergenic non-protein coding RNA 476
There are no transcripts associated with this gene.
View Full Product Details