Product Details

SNP ID
rs201489188
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35563316 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCCAGGCTAGGCCAGGAGGGCCTC[A/G]AAGTAGCTGACCAGTCACCTGCCCA
Phenotype
MIM: 611053
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM166B PubMed Links

Gene Details

Gene
FAM166B
Gene Name
family with sequence similarity 166 member B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099951.3 207 Nonsense Mutation CGA,TGA R46* NP_001093421.1
NM_001164310.2 207 Nonsense Mutation CGA,TGA R46* NP_001157782.1
NM_001287238.1 207 Nonsense Mutation CGA,TGA R46* NP_001274167.1
NM_001287239.1 207 Nonsense Mutation CGA,TGA R46* NP_001274168.1
XM_011518028.2 207 Nonsense Mutation CGA,TGA R41* XP_011516330.1
Gene
RUSC2
Gene Name
RUN and SH3 domain containing 2
There are no transcripts associated with this gene.

View Full Product Details