Product Details

SNP ID
rs200697165
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:114091922 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCCTCACTGCAGGGTGGCTGGCTG[C/T]GGCCACGTCGCAGGGAGCTGCCAAT
Phenotype
MIM: 611278
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KIF12 PubMed Links

Gene Details

Gene
KIF12
Gene Name
kinesin family member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138424.1 1980 Missense Mutation NP_612433.1
XM_005251683.4 1980 Missense Mutation XP_005251740.1
XM_006716947.2 1980 Missense Mutation XP_006717010.1
XM_011518190.2 1980 Missense Mutation XP_011516492.1
XM_011518191.2 1980 Missense Mutation XP_011516493.1
XM_011518192.2 1980 Intron XP_011516494.1
XM_011518193.2 1980 Intron XP_011516495.1
XM_017014240.1 1980 Missense Mutation XP_016869729.1

View Full Product Details