Product Details

SNP ID
rs200943052
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:135344161 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATCTAGCGGACAGCAGGGCTGCAG[A/C]GTGGACAGAACGGCCGGACGCCCAG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
C9orf62 PubMed Links

Gene Details

Gene
C9orf62
Gene Name
chromosome 9 open reading frame 62
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_173520.2 363 Missense Mutation AGA,AGC R71S NP_775791.1

View Full Product Details