Product Details
- SNP ID
-
rs200227684
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:977308 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCAGGGCCCCCGCCGCCGGGGGAC[A/G]CCGTCGCCGCCCCGCAGCCGCCGCC
- Phenotype
-
MIM: 602424
MIM: 614754
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
DMRT1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs148355218] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- DMRT1
- Gene Name
- doublesex and mab-3 related transcription factor 1
There are no transcripts associated with this gene.
- Gene
- DMRT3
- Gene Name
- doublesex and mab-3 related transcription factor 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_021240.3 |
341 |
Missense Mutation |
ACC,GCC |
T103A |
NP_067063.1 |
View Full Product Details