Product Details
- SNP ID
-
rs200267752
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:21239864 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TCAGGCTGTGGGTTTGAGACAGATT[A/G]CAGCCCAGAGAGCAGCTTGACTTGC
- Phenotype
-
MIM: 147579
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
IFNA14
PubMed Links
Gene Details
- Gene
- IFNA14
- Gene Name
- interferon alpha 14
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002172.2 |
116 |
Silent Mutation |
TGC,TGT |
C24C |
NP_002163.2 |
View Full Product Details