Product Details

SNP ID
rs200639946
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136362167 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGCTCGCCTCTGGCCGTCAGGA[C/T]CTCTTCGATATTTCTGGGGGGCAGG
Phenotype
MIM: 607212 MIM: 609491
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CARD9 PubMed Links

Gene Details

Gene
CARD9
Gene Name
caspase recruitment domain family member 9
There are no transcripts associated with this gene.

Gene
DNLZ
Gene Name
DNL-type zinc finger
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080849.2 479 Missense Mutation NP_001074318.1
Gene
GPSM1
Gene Name
G-protein signaling modulator 1
There are no transcripts associated with this gene.

View Full Product Details