Product Details

SNP ID
rs201011536
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:124873737 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCATGCAGCCTTGCGGAACTCTCC[C/T]GTCAACACCAAGAATCAAGCTGTGA
Phenotype
MIM: 602502
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ARPC5L PubMed Links

Gene Details

Gene
ARPC5L
Gene Name
actin related protein 2/3 complex subunit 5 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_030978.2 1459 Silent Mutation CCC,CCT P65P NP_112240.1
XM_011519067.1 1459 Silent Mutation CCC,CCT P13P XP_011517369.1
Gene
GOLGA1
Gene Name
golgin A1
There are no transcripts associated with this gene.

View Full Product Details