Product Details

SNP ID
rs200374086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:34379100 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTACCAGCAACGATAGTCGGGATA[G/T]GTCTCAGACACAAACTCAGGATGGA
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
C9orf24 PubMed Links

Gene Details

Gene
C9orf24
Gene Name
chromosome 9 open reading frame 24
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001252195.1 897 Silent Mutation ACA,ACC T157T NP_001239124.1
NM_032596.3 897 UTR 3 NP_115985.2
NM_147168.1 897 UTR 3 NP_671697.1
NM_147169.2 897 Silent Mutation ACA,ACC T104T NP_671698.1
XM_005251616.4 897 Silent Mutation ACA,ACC T292T XP_005251673.1
XM_005251618.4 897 Silent Mutation ACA,ACC T157T XP_005251675.1
XM_011518055.2 897 Silent Mutation ACA,ACC T157T XP_011516357.1
Gene
KIAA1161
Gene Name
KIAA1161
There are no transcripts associated with this gene.

View Full Product Details