Product Details

SNP ID
rs200744648
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:127732599 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCAGCACCGCGCGGGAGATGACC[A/G]GCTCCAGCTCCTGCAGGAGGATCTC
Phenotype
MIM: 604722 MIM: 608052
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SH2D3C PubMed Links

Gene Details

Gene
SH2D3C
Gene Name
SH2 domain containing 3C
There are no transcripts associated with this gene.

Gene
TOR2A
Gene Name
torsin family 2 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001085347.2 800 Missense Mutation CCG,CTG P229L NP_001078816.1
NM_001134430.2 800 Intron NP_001127902.1
NM_001134431.2 800 UTR 3 NP_001127903.1
NM_001252018.1 800 Missense Mutation CCG,CTG P67L NP_001238947.1
NM_001252021.1 800 Missense Mutation CCG,CTG P67L NP_001238950.1
NM_001252023.1 800 Intron NP_001238952.1
NM_130459.3 800 UTR 3 NP_569726.2
XM_011518554.1 800 Missense Mutation CCG,CTG P228L XP_011516856.1
Gene
TTC16
Gene Name
tetratricopeptide repeat domain 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317037.1 800 Intron NP_001303966.1
NM_144965.2 800 Intron NP_659402.1
XM_005251733.1 800 Intron XP_005251790.1
XM_005251734.1 800 Intron XP_005251791.1
XM_006716970.1 800 Intron XP_006717033.1
XM_006716971.1 800 Intron XP_006717034.1
XM_011518283.1 800 Intron XP_011516585.1
XM_011518284.1 800 Intron XP_011516586.1
XM_011518285.1 800 Intron XP_011516587.1
XM_011518288.1 800 Intron XP_011516590.1
XM_011518289.2 800 Intron XP_011516591.1
XM_011518290.1 800 Intron XP_011516592.1
XM_011518291.1 800 Intron XP_011516593.1
XM_011518292.1 800 Intron XP_011516594.1
XM_011518293.1 800 Intron XP_011516595.1
XM_011518294.2 800 Intron XP_011516596.1
XM_011518295.1 800 Intron XP_011516597.1
XM_011518297.1 800 Intron XP_011516599.1
XM_011518298.2 800 Intron XP_011516600.1

View Full Product Details