Product Details

SNP ID
rs200562136
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:98200253 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTGGCAGGGGGTGGGGGTTCCTCAC[C/T]GGCTGTTGGAGATGGTCTTGGTGAA
Phenotype
MIM: 602159 MIM: 609871
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CORO2A PubMed Links

Gene Details

Gene
CORO2A
Gene Name
coronin 2A
There are no transcripts associated with this gene.

Gene
TBC1D2
Gene Name
TBC1 domain family member 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001267571.1 2759 Missense Mutation NP_001254500.1
NM_001267572.1 2759 Missense Mutation NP_001254501.1
NM_018421.3 2759 Missense Mutation NP_060891.3
XM_011518843.1 2759 Missense Mutation XP_011517145.1
XM_011518844.2 2759 Intron XP_011517146.1
XM_017014899.1 2759 Missense Mutation XP_016870388.1

View Full Product Details