Product Details

SNP ID
rs201055450
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:97643341 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGAATCATTGAAAGCAAACAATT[A/G]TAATGAAGCCGTGTATTTGGTAAGT
Phenotype
MIM: 600469
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
NCBP1 PubMed Links

Gene Details

Gene
NCBP1
Gene Name
nuclear cap binding protein subunit 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002486.4 659 Missense Mutation TAT,TGT Y121C NP_002477.1
XM_011518715.2 659 Missense Mutation TAT,TGT Y121C XP_011517017.1
XM_017014735.1 659 Missense Mutation TAT,TGT Y54C XP_016870224.1
XM_017014736.1 659 UTR 5 XP_016870225.1
XM_017014737.1 659 UTR 5 XP_016870226.1
XM_017014738.1 659 UTR 5 XP_016870227.1
Gene
TSTD2
Gene Name
thiosulfate sulfurtransferase like domain containing 2
There are no transcripts associated with this gene.

View Full Product Details