Product Details

SNP ID
rs201362659
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33923245 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCTGCGATGATCCAGCATAGCCA[A/C]CTTTGGAGTAGTCTCCTGCTGCTGT
Phenotype
MIM: 612506
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
UBAP2 PubMed Links

Gene Details

Gene
UBAP2
Gene Name
ubiquitin associated protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282529.1 3058 Missense Mutation GGT,GTT G715V NP_001269458.1
NM_001282530.1 3058 Missense Mutation GGT,GTT G221V NP_001269459.1
NM_018449.3 3058 Missense Mutation GGT,GTT G982V NP_060919.3
Gene
UBE2R2
Gene Name
ubiquitin conjugating enzyme E2 R2
There are no transcripts associated with this gene.

View Full Product Details