Product Details

SNP ID
rs200501115
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:68780220 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGCTGCCTCCGGGTACGGGCGGGA[G/T]CCCGGCGGAGGGCGGTGGCAGCGGC
Phenotype
MIM: 602745
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FAM122A PubMed Links

Gene Details

Gene
FAM122A
Gene Name
family with sequence similarity 122A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138333.4 215 Missense Mutation AGC,ATC S61I NP_612206.4
Gene
PIP5K1B
Gene Name
phosphatidylinositol-4-phosphate 5-kinase type 1 beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001278253.1 215 Intron NP_001265182.1
NM_003558.3 215 Intron NP_003549.1
XM_005252261.3 215 Intron XP_005252318.1
XM_005252262.4 215 Intron XP_005252319.1
XM_006717300.2 215 Intron XP_006717363.1
XM_006717301.1 215 Intron XP_006717364.1
XM_011519082.2 215 Intron XP_011517384.1
XM_011519083.2 215 Intron XP_011517385.1
XM_011519084.2 215 Intron XP_011517386.1
XM_017015188.1 215 Intron XP_016870677.1
XM_017015189.1 215 Intron XP_016870678.1
XM_017015190.1 215 Intron XP_016870679.1
XM_017015191.1 215 Intron XP_016870680.1

View Full Product Details