Product Details

SNP ID
rs200195618
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:35616139 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGTAGTGCTTCCTGACTCTGCGC[C/T]AGCTCTCTCCTGGACCCCTGCAGAT
Phenotype
MIM: 107272 MIM: 601782
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CD72 PubMed Links

Gene Details

Gene
CD72
Gene Name
CD72 molecule
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001782.2 989 Silent Mutation CTA,CTG L164L NP_001773.1
XM_006716893.2 989 Silent Mutation CTA,CTG L164L XP_006716956.1
XM_006716894.3 989 Silent Mutation CTA,CTG L160L XP_006716957.1
XM_011518074.2 989 Silent Mutation CTA,CTG L160L XP_011516376.1
XM_017015351.1 989 Silent Mutation CTA,CTG L160L XP_016870840.1
Gene
MIR4667
Gene Name
microRNA 4667
There are no transcripts associated with this gene.

Gene
TESK1
Gene Name
testis-specific kinase 1
There are no transcripts associated with this gene.

View Full Product Details