Product Details

SNP ID
rs200378599
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133536723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTGCTTCCTAGGATGGATGGCA[C/G]ATGGCAATGTTCCTGCTGGGCCTGG
Phenotype
MIM: 612277
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ADAMTSL2 PubMed Links

Gene Details

Gene
ADAMTSL2
Gene Name
ADAMTS like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145320.1 226 Missense Mutation ACA,AGA T4R NP_001138792.1
NM_014694.3 226 Missense Mutation ACA,AGA T4R NP_055509.2
XM_005272237.3 226 Missense Mutation ACA,AGA T113R XP_005272294.1
XM_005272238.3 226 Missense Mutation CAC,CAG H63Q XP_005272295.1
XM_005272239.2 226 Missense Mutation ACA,AGA T4R XP_005272296.1
XM_011519241.2 226 Missense Mutation ACA,AGA T113R XP_011517543.2
XM_011519242.2 226 Silent Mutation CAC,CAG H63Q XP_011517544.1

View Full Product Details