Product Details

SNP ID
rs200831920
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:122526449 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGGCTATAGATAAAGGGGTTCAAC[A/G]TGGGAGTCACTATGGTGTACATTGC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
OR1J4 PubMed Links

Gene Details

Gene
OR1J4
Gene Name
olfactory receptor family 1 subfamily J member 4
There are no transcripts associated with this gene.

Gene
OR1N1
Gene Name
olfactory receptor family 1 subfamily N member 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012363.1 Intron NP_036495.1

View Full Product Details