Product Details

SNP ID
rs200852198
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:137214068 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCACTGAGATCCCTCCCGGAGTCC[C/G]GTGAGTGTGGGCCCCGGGTCACCCA
Phenotype
MIM: 606073 MIM: 616499
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NDOR1 PubMed Links

Gene Details

Gene
NDOR1
Gene Name
NADPH dependent diflavin oxidoreductase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001144026.2 1782 Missense Mutation CCC,CGC P171R NP_001137498.1
NM_001144027.2 1782 Intron NP_001137499.1
NM_001144028.2 1782 Missense Mutation CCC,CGC P171R NP_001137500.1
NM_014434.3 1782 Missense Mutation CCC,CGC P171R NP_055249.1
XM_011518545.1 1782 Intron XP_011516847.1
XM_011518547.1 1782 Missense Mutation CCC,CGC P102R XP_011516849.1
XM_011518548.2 1782 UTR 5 XP_011516850.1
XM_017014626.1 1782 Missense Mutation CCC,CGC P171R XP_016870115.1
XM_017014627.1 1782 Missense Mutation CCC,CGC P171R XP_016870116.1
Gene
RNF208
Gene Name
ring finger protein 208
There are no transcripts associated with this gene.

Gene
TMEM203
Gene Name
transmembrane protein 203
There are no transcripts associated with this gene.

View Full Product Details