Product Details
- SNP ID
-
rs201159012
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:74724716 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CTCAAGTCCAAAGGTGGAATTTATC[A/C]TTTCAGGGGAATAGTCATTTCTTTT
- Phenotype
-
MIM: 607009
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TRPM6
PubMed Links
Gene Details
- Gene
- TRPM6
- Gene Name
- transient receptor potential cation channel subfamily M member 6
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001177310.1 |
5928 |
Missense Mutation |
AGG,ATG |
R1984M |
NP_001170781.1 |
NM_001177311.1 |
5928 |
Missense Mutation |
AGG,ATG |
R1984M |
NP_001170782.1 |
NM_017662.4 |
5928 |
Missense Mutation |
AGG,ATG |
R1989M |
NP_060132.3 |
XM_011518244.1 |
5928 |
Missense Mutation |
AGG,ATG |
R1988M |
XP_011516546.1 |
XM_011518245.1 |
5928 |
Missense Mutation |
AGG,ATG |
R1958M |
XP_011516547.1 |
XM_011518246.1 |
5928 |
Missense Mutation |
AGG,ATG |
R1948M |
XP_011516548.1 |
XM_011518247.1 |
5928 |
Missense Mutation |
AGG,ATG |
R1946M |
XP_011516549.1 |
XM_011518248.1 |
5928 |
Missense Mutation |
AGG,ATG |
R1942M |
XP_011516550.1 |
XM_011518249.1 |
5928 |
Missense Mutation |
AGG,ATG |
R1911M |
XP_011516551.1 |
XM_011518250.1 |
5928 |
Missense Mutation |
AGG,ATG |
R1897M |
XP_011516552.1 |
XM_011518251.2 |
5928 |
Missense Mutation |
AGG,ATG |
R1746M |
XP_011516553.1 |
XM_011518252.2 |
5928 |
Intron |
|
|
XP_011516554.1 |
XM_011518255.2 |
5928 |
Intron |
|
|
XP_011516557.1 |
XM_017014287.1 |
5928 |
Missense Mutation |
AGG,ATG |
R1868M |
XP_016869776.1 |
XM_017014288.1 |
5928 |
Missense Mutation |
AGG,ATG |
R1819M |
XP_016869777.1 |
XM_017014289.1 |
5928 |
Intron |
|
|
XP_016869778.1 |
View Full Product Details