Product Details

SNP ID
rs200276837
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136941300 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCAGGAAGATGAAGAAGCACTCGT[C/T]GTTGGGCGTGTAGGCGCGGTGCGCA
Phenotype
MIM: 120930 MIM: 609072 MIM: 612905
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C8G PubMed Links

Gene Details

Gene
C8G
Gene Name
complement component 8, gamma polypeptide
There are no transcripts associated with this gene.

Gene
FBXW5
Gene Name
F-box and WD repeat domain containing 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018998.3 1588 Missense Mutation AAC,GAC N470D NP_061871.1
XM_005266089.2 1588 Missense Mutation AAC,GAC N480D XP_005266146.2
XM_011518789.1 1588 Missense Mutation AAC,GAC N462D XP_011517091.1
XM_017014812.1 1588 Missense Mutation AAC,GAC N452D XP_016870301.1
Gene
LCN12
Gene Name
lipocalin 12
There are no transcripts associated with this gene.

View Full Product Details