Product Details

SNP ID
rs200621653
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:1465998 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGAGGAACACGGCCCACATGGC[C/T]ACGCTGGGGAGCATCGTCCACTTGA
Phenotype
MIM: 300525
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
P2RY8 PubMed Links

Gene Details

Gene
P2RY8
Gene Name
purinergic receptor P2Y8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_178129.4 926 Silent Mutation GTA,GTG V187V NP_835230.1
XM_005274429.3 926 Silent Mutation GTA,GTG V187V XP_005274486.1
XM_006724443.3 926 Silent Mutation GTA,GTG V376V XP_006724506.2
XM_011546178.2 926 Silent Mutation GTA,GTG V187V XP_011544480.1
XM_011546179.2 926 Silent Mutation GTA,GTG V187V XP_011544481.1

View Full Product Details