Product Details

SNP ID
rs199547236
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:36169953 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGAGCTCAAAGAGGTATCAGAC[A/G]TAGCTTACTCTGGCATTCGGGACAA
Phenotype
MIM: 603960 MIM: 607141
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCIN PubMed Links

Gene Details

Gene
CCIN
Gene Name
calicin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005893.2 562 Missense Mutation ATA,GTA I151V NP_005884.2
Gene
GLIPR2
Gene Name
GLI pathogenesis related 2
There are no transcripts associated with this gene.

View Full Product Details