Product Details

SNP ID
rs199835146
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:120905184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAGGACGCTGAGCTTAGGTCAGGCC[A/G]GAAGGCGTCAATGGCGTGCTCACGG
Phenotype
MIM: 609740 MIM: 601711
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PHF19 PubMed Links

Gene Details

Gene
PHF19
Gene Name
PHD finger protein 19
There are no transcripts associated with this gene.

Gene
TRAF1
Gene Name
TNF receptor associated factor 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001190945.1 1512 Silent Mutation NP_001177874.1
NM_001190947.1 1512 Silent Mutation NP_001177876.1
NM_005658.4 1512 Silent Mutation NP_005649.1

View Full Product Details