Product Details

SNP ID
rs199918911
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133364945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTTTGGGGGAGCTCTCACGCATGGT[A/G]GGGACGCCCGCAAACATGCTCTTCC
Phenotype
MIM: 185620 MIM: 185630 MIM: 185660
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SURF1 PubMed Links

Gene Details

Gene
SURF1
Gene Name
surfeit 1
There are no transcripts associated with this gene.

Gene
SURF2
Gene Name
surfeit 2
There are no transcripts associated with this gene.

Gene
SURF4
Gene Name
surfeit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001280788.1 635 Silent Mutation CCC,CCT P131P NP_001267717.1
NM_001280789.1 635 Silent Mutation CCC,CCT P146P NP_001267718.1
NM_001280790.1 635 Silent Mutation CCC,CCT P103P NP_001267719.1
NM_001280791.1 635 Silent Mutation CCC,CCT P103P NP_001267720.1
NM_001280792.1 635 Intron NP_001267721.1
NM_001280793.1 635 Silent Mutation CCC,CCT P29P NP_001267722.1
NM_033161.3 635 Silent Mutation CCC,CCT P146P NP_149351.1
XM_011518943.1 635 Silent Mutation CCC,CCT P137P XP_011517245.1
XM_011518944.2 635 Silent Mutation CCC,CCT P137P XP_011517246.1
XM_011518945.2 635 Intron XP_011517247.1
XM_017015062.1 635 Intron XP_016870551.1

View Full Product Details