Product Details

SNP ID
rs199856885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:5300197 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCTCTTTTTTCGAGAATGAGTATC[C/T]AAGCCTAAGTATTTTAATTCTGAAG
Phenotype
MIM: 179730 MIM: 179740
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
RLN1 PubMed Links

Gene Details

Gene
RLN1
Gene Name
relaxin 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006911.3 591 Intron NP_008842.1
Gene
RLN2
Gene Name
relaxin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005059.3 591 UTR 3 NP_005050.2
NM_134441.2 591 Silent Mutation TTA,TTG L153L NP_604390.1
XM_017015011.1 591 Silent Mutation TTA,TTG L137L XP_016870500.1

View Full Product Details