Product Details
- SNP ID
-
rs199551289
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:32784631 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGGAAGGAGAATGCCAGAGCCTC[C/G]TCCAGAATGGGCATCAGGAGGAGAC
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
TMEM215
PubMed Links
Gene Details
- Gene
- TMEM215
- Gene Name
- transmembrane protein 215
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_212558.2 |
813 |
Missense Mutation |
CTC,GTC |
L150V |
NP_997723.2 |
View Full Product Details