Product Details

SNP ID
rs199654769
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37592511 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCTCCTCCGGGTCCAGCTTCGGCG[A/C]GAGGCCCTCAATCCGGAACATCGCG
Phenotype
MIM: 616169
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
TOMM5 PubMed Links

Gene Details

Gene
TOMM5
Gene Name
translocase of outer mitochondrial membrane 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001790.2 129 Missense Mutation GCG,TCG A8S NP_001001790.1
NM_001134484.1 129 Missense Mutation GCG,TCG A8S NP_001127956.1
NM_001134485.1 129 Missense Mutation GCG,TCG A8S NP_001127957.1

View Full Product Details