Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001131015.1 | 2536 | Missense Mutation | NP_001124487.1 | ||
NM_001131016.1 | 2536 | Missense Mutation | NP_001124488.1 | ||
NM_001131017.1 | 2536 | Missense Mutation | NP_001124489.1 | ||
NM_001131018.1 | 2536 | Missense Mutation | NP_001124490.1 | ||
NM_001257975.1 | 2536 | Missense Mutation | NP_001244904.1 | ||
NM_001257976.1 | 2536 | Missense Mutation | NP_001244905.1 | ||
NM_012127.2 | 2536 | Missense Mutation | NP_036259.2 | ||
XM_005251888.3 | 2536 | Missense Mutation | XP_005251945.2 | ||
XM_005251891.3 | 2536 | Missense Mutation | XP_005251948.2 | ||
XM_005251892.3 | 2536 | Missense Mutation | XP_005251949.2 | ||
XM_005251893.4 | 2536 | Missense Mutation | XP_005251950.4 | ||
XM_006717039.3 | 2536 | Missense Mutation | XP_006717102.2 | ||
XM_011518481.1 | 2536 | Missense Mutation | XP_011516783.1 | ||
XM_011518482.2 | 2536 | Missense Mutation | XP_011516784.2 | ||
XM_011518483.1 | 2536 | Missense Mutation | XP_011516785.1 | ||
XM_017014594.1 | 2536 | Missense Mutation | XP_016870083.1 | ||
XM_017014595.1 | 2536 | Missense Mutation | XP_016870084.1 | ||
XM_017014596.1 | 2536 | Missense Mutation | XP_016870085.1 |