Product Details

SNP ID
rs201532959
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:114092348 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTTCTTCACCTCTGAGCCCTGGTG[A/T]TGTCTTCGGGGGCCTCACAGGCAGG
Phenotype
MIM: 611278
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KIF12 PubMed Links

Gene Details

Gene
KIF12
Gene Name
kinesin family member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138424.1 1866 Missense Mutation ACA,TCA T463S NP_612433.1
XM_005251683.4 1866 Missense Mutation ACA,TCA T601S XP_005251740.1
XM_006716947.2 1866 Missense Mutation ACA,TCA T567S XP_006717010.1
XM_011518190.2 1866 Missense Mutation ACA,TCA T659S XP_011516492.1
XM_011518191.2 1866 Missense Mutation ACA,TCA T659S XP_011516493.1
XM_011518192.2 1866 Intron XP_011516494.1
XM_011518193.2 1866 Intron XP_011516495.1
XM_017014240.1 1866 Missense Mutation ACA,TCA T336S XP_016869729.1

View Full Product Details