Product Details

SNP ID
rs201593266
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:37486801 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACCCCAGGAAGAGGAATCAACGG[A/G]TCCTGGTAAGTGAAGCAGTTGCCAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
POLR1E PubMed Links

Gene Details

Gene
POLR1E
Gene Name
RNA polymerase I subunit E
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282766.1 288 UTR 5 NP_001269695.1
NM_022490.2 288 Missense Mutation ATC,GTC I59V NP_071935.1
XM_005251547.2 288 Missense Mutation ATC,GTC I121V XP_005251604.1

View Full Product Details