Product Details
- SNP ID
-
rs201555289
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:114403960 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCTCCTTGCTACTCCTGCTCTTG[A/G]TGGACACCGACTGCCTTCCTCGGCC
- Phenotype
-
MIM: 605729
MIM: 607928
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
AKNA
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2274159] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- AKNA
- Gene Name
- AT-hook transcription factor
There are no transcripts associated with this gene.
- Gene
- WHRN
- Gene Name
- whirlin
View Full Product Details