Product Details
- SNP ID
-
rs201989691
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:129739614 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCAAGGAGGCATCAGCTGCTGGTCA[C/T]AGGTGGCGGGCCGCTTCCCAGAGGA
- Phenotype
-
MIM: 605172
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
PTGES
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2302821] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- PTGES
- Gene Name
- prostaglandin E synthase
View Full Product Details