Product Details

SNP ID
rs201538126
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:108939240 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTTGCACCTCTCTGTAAGATTTA[C/T]TGCTCAGATGGTGAAGAATATACTG
Phenotype
MIM: 604785 MIM: 603722
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTNNAL1 PubMed Links

Gene Details

Gene
CTNNAL1
Gene Name
catenin alpha like 1
There are no transcripts associated with this gene.

Gene
FAM206A
Gene Name
family with sequence similarity 206 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_017832.3 400 Silent Mutation TAC,TAT Y102Y NP_060302.1
XM_011518816.2 400 Intron XP_011517118.1
Gene
IKBKAP
Gene Name
inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase complex-associated protein
There are no transcripts associated with this gene.

View Full Product Details