Product Details

SNP ID
rs201725177
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:128276332 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTGCACCTGAGAGGTCGCTCTCCG[A/G]CTCCCGCGCTGGACCCTCTTGCGCC
Phenotype
MIM: 602580 MIM: 616528
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GOLGA2 PubMed Links

Gene Details

Gene
GOLGA2
Gene Name
golgin A2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004486.4 187 Intron NP_004477.3
XM_005251930.1 187 Intron XP_005251987.1
XM_005251931.1 187 Intron XP_005251988.1
XM_005251932.1 187 Intron XP_005251989.1
Gene
SWI5
Gene Name
SWI5 homologous recombination repair protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040011.1 187 Missense Mutation ACT,GCT T63A NP_001035100.1
NM_001318089.1 187 Missense Mutation ACT,GCT T63A NP_001305018.1
NM_001318092.1 187 Intron NP_001305021.1
XM_011518652.2 187 Missense Mutation ACT,GCT T63A XP_011516954.1

View Full Product Details