Product Details

SNP ID
rs201870375
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:135624226 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGAAGTCTAAAATATTATGGGGTGT[G/T]CCACCAACCATCAAGTTGCCATAAT
Phenotype
MIM: 613699
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GLT6D1 PubMed Links

Gene Details

Gene
GLT6D1
Gene Name
glycosyltransferase 6 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182974.2 1280 Silent Mutation GGA,GGC G234G NP_892019.2
XM_011518633.1 1280 Silent Mutation GGA,GGC G270G XP_011516935.1
XM_011518634.1 1280 Silent Mutation GGA,GGC G270G XP_011516936.1
XM_011518635.1 1280 Silent Mutation GGA,GGC G270G XP_011516937.1
XM_011518636.1 1280 Silent Mutation GGA,GGC G270G XP_011516938.1
XM_011518637.1 1280 Silent Mutation GGA,GGC G234G XP_011516939.1
XM_011518638.2 1280 Silent Mutation GGA,GGC G202G XP_011516940.1
Gene
LOC102723971
Gene Name
lipocalin 1-like
There are no transcripts associated with this gene.

View Full Product Details