Product Details

SNP ID
rs201851161
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:136412041 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACGGTTTAGTAGTGGTGGTGCCT[A/G]TCCCAACATCCCCCTCTCTTCTCCC
Phenotype
MIM: 613036
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PMPCA PubMed Links

Gene Details

Gene
PMPCA
Gene Name
peptidase, mitochondrial processing alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282944.1 127 Intron NP_001269873.1
NM_001282946.1 127 Intron NP_001269875.1
NM_015160.2 127 Intron NP_055975.1
XM_005266059.4 127 Missense Mutation TAT,TGT Y39C XP_005266116.1
XM_011518417.2 127 Intron XP_011516719.1
XM_017014543.1 127 Intron XP_016870032.1
Gene
SDCCAG3
Gene Name
serologically defined colon cancer antigen 3
There are no transcripts associated with this gene.

View Full Product Details