Product Details

SNP ID
rs201645553
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:107487075 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTGAGATGGGAACTCTTTGTGTAG[C/G]TTTTGCCGCAGCCCGCGTAATCACA
Phenotype
MIM: 602253
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
KLF4 PubMed Links

Gene Details

Gene
KLF4
Gene Name
Kruppel like factor 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001314052.1 1765 Missense Mutation ACC,AGC T440S NP_001300981.1
NM_004235.5 1765 Missense Mutation ACC,AGC T406S NP_004226.3

View Full Product Details